Assay Details
| Gene Symbol: Nupl1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_170591.1 | NP_733479.1 | 7 - 8 | 854 | 84 | |
| GenBank mRNA | AK006062.1 | - | 7 - 8 | 956 | 84 | |
| AK029688.1 | - | 908 | 84 | |||
| AK078279.1 | - | 7 - 8 | 902 | 84 | ||
| AK086412.1 | - | 7 - 8 | 922 | 84 | ||
| AK129134.1 | - | 7 - 8 | 852 | 84 | ||
| AK133721.1 | - | 7 - 8 | 865 | 84 | ||
| AK139895.1 | - | 7 - 8 | 985 | 84 | ||
| AK146477.1 | - | 7 - 8 | 854 | 84 | ||
| BC026743.1 | - | 7 - 8 | 885 | 3964978 | 84 | |
More Information
Set Membership: |
|

Nupl1
Genomic Map