Assay Details
| Gene Symbol: Mipep | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027436.3 | NP_081712.2 | 5 - 6 | 721 | 115 | |
| NR_040642.1 | 5 - 6 | 721 | 115 | |||
| GenBank mRNA | AK017491.1 | - | 5 - 6 | 697 | 115 | |
| AK038479.1 | - | 5 - 6 | 715 | 115 | ||
| AK044460.1 | - | 5 - 6 | 706 | 115 | ||
| AK076613.1 | - | 5 - 6 | 649 | 115 | ||
| AK146428.1 | - | 6 - 7 | 712 | 115 | ||
| BC110358.1 | - | 5 - 6 | 699 | 5322591 | 115 | |
| BC144798.1 | - | 5 - 6 | 702 | 115 | ||
| BC145711.1 | - | 5 - 6 | 702 | 40131014 | 115 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mipep
Genomic Map