Assay Details
| Gene Symbol: Peli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033602.2 | NP_291080.2 | 1 - 2 | 363 | 72 | |
| GenBank mRNA | AK082342.1 | - | 1 - 2 | 319 | 72 | |
More Information
Updated Mapping Information
Set Membership: |
|

Peli2
Genomic Map