Assay Details
| Gene Symbol: Peli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033602.2 | NP_291080.2 | 2 - 3 | 502 | 82 | |
| GenBank mRNA | AF302504.1 | - | 2 - 3 | 431 | 82 | |
| AK030564.1 | - | 2 - 3 | 549 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Peli2
Genomic Map