Assay Details
| Gene Symbol: Peli2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033602.2 | NP_291080.2 | 4 - 5 | 797 | 81 | |
| GenBank mRNA | AF302504.1 | - | 4 - 5 | 726 | 81 | |
| AK030564.1 | - | 4 - 5 | 844 | 81 | ||
| AK033815.1 | - | 1 - 2 | 103 | 81 | ||
| AK045515.1 | - | 3 - 4 | 670 | 81 | ||
| BC027062.1 | - | 3 - 4 | 636 | 5355240 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Peli2
Genomic Map