Assay Details
| Gene Symbol: Piwil2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021308.1 | NP_067283.1 | 20 - 21 | 2593 | 75 | |
| GenBank mRNA | AB032605.1 | - | 20 - 21 | 2593 | 75 | |
| AF285586.1 | - | 11 - 12 | 1322 | 75 | ||
| AK030116.1 | - | 20 - 21 | 2537 | 75 | ||
| AK163647.1 | - | 7 - 8 | 848 | 75 | ||
| BC138444.1 | - | 20 - 21 | 2581 | 8861464 | 75 | |
| BC145717.1 | - | 20 - 21 | 2581 | 40131020 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Piwil2
Genomic Map