Assay Details
| Gene Symbol: Clasrp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016680.5 | NP_057889.3 | 15 - 16 | 1781 | 90 | |
| GenBank mRNA | AB080582.1 | - | 14 - 15 | 1644 | 90 | |
| AF325456.1 | - | 5 - 6 | 1627 | 90 | ||
| BC010728.1 | - | 15 - 16 | 1733 | 90 | ||
| BC060092.1 | - | 14 - 15 | 4149 | 90 | ||
More Information
Set Membership: |
|

Clasrp
Genomic Map