Assay Details
| Gene Symbol: Rtn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013648.6 | NP_038676.1 | 4 - 5 | 1023 | 79 | |
| GenBank mRNA | AF038538.1 | - | 4 - 5 | 979 | 79 | |
| AK144271.1 | - | 4 - 5 | 986 | 79 | ||
| BC031370.1 | - | 4 - 5 | 980 | 5359358 | 79 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rtn2
Genomic Map