Assay Details
| Gene Symbol: Sycp1-ps1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_024208.1 | 1 | 2200 | 121 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | pregnancy-specific glycoprotein 22 | Psg22 |
AA408604, cea9 |
-59.1kb | Sense | Chr. 7 - 18718090 - 18727248 on NCBI Build GRCm38 |
| Downstream | pregnancy specific glycoprotein 17 | Psg17 |
Cea-2, Cea2, mmCGM5 |
24.5kb | Anti-sense | Chr. 7 - 18813937 - 18821591 on NCBI Build GRCm38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sycp1-ps1
Genomic Map