Assay Details
| Gene Symbol: Ephx2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001271402.1 | NP_001258331.1 | 16 - 17 | 1610 | 73 | |
| NM_001271403.1 | NP_001258332.1 | 15 - 16 | 1455 | 73 | ||
| NM_001271421.1 | NP_001258350.1 | 15 - 16 | 1505 | 73 | ||
| NM_007940.4 | NP_031966.2 | 16 - 17 | 1530 | 73 | ||
| GenBank mRNA | AK002415.1 | - | 16 - 17 | 1580 | 73 | |
| AK142714.1 | - | 16 - 17 | 1503 | 73 | ||
| AY098585.1 | - | 15 - 16 | 1469 | 73 | ||
| BC015087.1 | - | 16 - 17 | 1506 | 4218917 | 73 | |
| EF597241.1 | - | 15 - 16 | 1505 | 73 | ||
| L05781.1 | - | 16 - 17 | 1443 | 73 | ||
| Z37107.1 | - | 16 - 17 | 1460 | 73 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ephx2
Genomic Map