Assay Details
| Gene Symbol: Dcaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199009.1 | NP_001185938.1 | 11 - 12 | 1364 | 93 | |
| NM_133734.3 | NP_598495.1 | 11 - 12 | 1269 | 93 | ||
| NR_037572.1 | 11 - 12 | 1381 | 93 | |||
| GenBank mRNA | AK033540.1 | - | 11 - 12 | 1267 | 93 | |
| AK150183.1 | - | 11 - 12 | 1356 | 93 | ||
| AK152395.1 | - | 10 - 11 | 1322 | 93 | ||
| AK158784.1 | - | 11 - 12 | 1349 | 93 | ||
| AK170657.1 | - | 11 - 12 | 1267 | 93 | ||
| BC008545.1 | - | 10 - 11 | 1162 | 3497641 | 93 | |
| BC021305.1 | - | 8 - 9 | 3178 | 93 | ||
| BC037001.1 | - | 10 - 11 | 1293 | 5051353 | 93 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK003028.1 | - | Chr. - - [-] on Build | 93 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf11
Genomic Map