Assay Details
| Gene Symbol: Dcaf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199009.1 | NP_001185938.1 | 9 - 10 | 1113 | 91 | |
| NM_133734.3 | NP_598495.1 | 9 - 10 | 1018 | 91 | ||
| NR_037572.1 | 9 - 10 | 1130 | 91 | |||
| GenBank mRNA | AK033540.1 | - | 9 - 10 | 1016 | 91 | |
| AK150183.1 | - | 9 - 10 | 1105 | 91 | ||
| AK152395.1 | - | 8 - 9 | 1071 | 91 | ||
| AK158784.1 | - | 9 - 10 | 1098 | 91 | ||
| AK170657.1 | - | 9 - 10 | 1016 | 91 | ||
| AK179635.1 | - | 214 | 91 | |||
| BC008545.1 | - | 8 - 9 | 911 | 3497641 | 91 | |
| BC037001.1 | - | 8 - 9 | 1042 | 5051353 | 91 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | AK003028.1 | - | Chr. - - [-] on Build | 91 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Dcaf11
Genomic Map