Assay Details
| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194346.2 | NP_919327.2 | 11 - 12 | 2208 | 75 | |
| GenBank mRNA | AK156320.1 | - | 11 - 12 | 2291 | 75 | |
| AY283085.1 | - | 11 - 12 | 2117 | 75 | ||
| BC057595.1 | - | 11 - 12 | 2208 | 6811051 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf31
Genomic Map