Assay Details
| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194346.2 | NP_919327.2 | 12 - 13 | 2374 | 86 | |
| GenBank mRNA | AK131160.1 | - | 5 - 6 | 3933 | 86 | |
| AK156320.1 | - | 12 - 13 | 2457 | 86 | ||
| AK216380.1 | - | 1 - 2 | 120 | 86 | ||
| AY283085.1 | - | 12 - 13 | 2283 | 86 | ||
| BC057595.1 | - | 12 - 13 | 2374 | 6811051 | 86 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf31
Genomic Map