Assay Details
| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194346.2 | NP_919327.2 | 15 - 16 | 2689 | 61 | |
| GenBank mRNA | AK131160.1 | - | 8 - 9 | 4248 | 61 | |
| AK156320.1 | - | 15 - 16 | 2772 | 61 | ||
| AY283085.1 | - | 15 - 16 | 2598 | 61 | ||
| BC057595.1 | - | 15 - 16 | 2689 | 6811051 | 61 | |
More Information
Set Membership: |
|

Rnf31
Genomic Map