Assay Details
| Gene Symbol: Rnf31 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_194346.2 | NP_919327.2 | 3 - 4 | 581 | 57 | |
| GenBank mRNA | AK156320.1 | - | 3 - 4 | 664 | 57 | |
| AK157962.1 | - | 3 - 4 | 627 | 57 | ||
| AK196715.1 | - | 1 - 2 | 145 | 57 | ||
| AY283085.1 | - | 3 - 4 | 487 | 57 | ||
| BC057595.1 | - | 3 - 4 | 581 | 6811051 | 57 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnf31
Genomic Map