Assay Details
| Gene Symbol: Hsd11b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001044751.1 | NP_001038216.1 | 2 - 3 | 323 | 64 | |
| NM_008288.2 | NP_032314.2 | 3 - 4 | 349 | 64 | ||
| GenBank mRNA | AI099306.1 | - | 2 - 3 | 326 | 64 | |
| AK149518.1 | - | 2 - 3 | 321 | 64 | ||
| AK165212.1 | - | 2 - 3 | 321 | 64 | ||
| AK167419.1 | - | 1 - 2 | 273 | 64 | ||
| BC132364.1 | - | 3 - 4 | 289 | 40130641 | 64 | |
| BC138780.1 | - | 3 - 4 | 289 | 8861802 | 64 | |
| BC145415.1 | - | 3 - 4 | 289 | 9053973 | 64 | |
| DQ089001.1 | - | 2 - 3 | 225 | 64 | ||
| S75207.1 | - | 2 - 3 | 270 | 64 | ||
| X83202.1 | - | 3 - 4 | 352 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Hsd11b1
Genomic Map