| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200000259.mm.1 |
NM_001271753.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000259.mm.1 |
NM_007872.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000259.mm.1 |
NM_007872.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000259.mm.1 |
NM_153743.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000259.mm.1 |
TC1200000016.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200000258.mm.1 |
NM_001271753.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000258.mm.1 |
NM_007872.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000258.mm.1 |
NM_007872.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000258.mm.1 |
NM_153743.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000258.mm.1 |
TC1200000016.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200000257.mm.1 |
NM_001271753.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000257.mm.1 |
NM_007872.4.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000257.mm.1 |
NM_007872.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000257.mm.1 |
NM_153743.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200000257.mm.1 |
TC1200000016.mm.1 |
MTA TC |