| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0100030487.mm.1 |
NM_001278220.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030487.mm.1 |
TC0100003721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030481.mm.1 |
NM_001278218.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030481.mm.1 |
NM_001278219.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030481.mm.1 |
NM_001278220.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030481.mm.1 |
NM_026866.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030481.mm.1 |
TC0100003721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030485.mm.1 |
NM_001278220.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030485.mm.1 |
TC0100003721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030483.mm.1 |
NM_001278218.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030483.mm.1 |
NM_001278219.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030483.mm.1 |
NM_001278220.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030483.mm.1 |
NM_026866.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030483.mm.1 |
TC0100003721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030486.mm.1 |
NM_001278220.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030486.mm.1 |
TC0100003721.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0100030482.mm.1 |
NM_001278218.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030482.mm.1 |
NM_001278219.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030482.mm.1 |
NM_001278220.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030482.mm.1 |
NM_026866.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0100030482.mm.1 |
TC0100003721.mm.1 |
MTA TC |