Assay Details
| Gene Symbol: Esrrg | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243792.1 | NP_001230721.1 | 5 - 6 | 935 | 61 | |
| NM_011935.3 | NP_036065.1 | 4 - 5 | 873 | 61 | ||
| GenBank mRNA | AB093266.1 | - | 5 - 6 | 954 | 61 | |
| AB221580.1 | - | 4 - 5 | 707 | 61 | ||
| AF117254.1 | - | 4 - 5 | 770 | 61 | ||
| AK052731.1 | - | 5 - 6 | 914 | 61 | ||
| AK140676.1 | - | 2 - 3 | 222 | 61 | ||
| BC082324.1 | - | 4 - 5 | 841 | 6850748 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Esrrg
Genomic Map