Assay Details
| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035061.1 | - | 3 - 4 | 848 | 94 | |
| AK089158.1 | - | 3 - 4 | 801 | 94 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Mettl3| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK035061.1 | - | 3 - 4 | 848 | 94 | |
| AK089158.1 | - | 3 - 4 | 801 | 94 | ||
Set Membership: |
|