Assay Details
| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019721.2 | NP_062695.2 | 8 - 9 | 1587 | 81 | |
| GenBank mRNA | AF135789.1 | - | 6 - 7 | 1150 | 81 | |
| AK035061.1 | - | 7 - 8 | 1405 | 81 | ||
| AK085189.1 | - | 8 - 9 | 1574 | 81 | ||
More Information
Set Membership: |
|

Mettl3
Genomic Map