Assay Details
| Gene Symbol: Supt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033618.3 | NP_291096.2 | 10 - 11 | 1322 | 164 | |
| GenBank mRNA | AK146529.1 | - | 10 - 11 | 1303 | 164 | |
| AK148506.1 | - | 10 - 11 | 1275 | 164 | ||
| AK169561.1 | - | 10 - 11 | 1324 | 164 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt16
Genomic Map