Assay Details
| Gene Symbol: Supt16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_033618.3 | NP_291096.2 | 7 - 8 | 1036 | 128 | |
| GenBank mRNA | AF323667.1 | - | 7 - 8 | 998 | 128 | |
| AK146529.1 | - | 7 - 8 | 1017 | 128 | ||
| AK148506.1 | - | 7 - 8 | 989 | 128 | ||
| AK169561.1 | - | 7 - 8 | 1038 | 128 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Supt16
Genomic Map