Assay Details
| Gene Symbol: Ndrg2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145959.1 | NP_001139431.1 | 3 - 4 | 350 | 112 | |
| NM_013864.2 | NP_038892.1 | 4 - 5 | 392 | 112 | ||
| GenBank mRNA | AB033921.1 | - | 4 - 5 | 255 | 112 | |
| AK154597.1 | - | 4 - 5 | 260 | 112 | ||
| AK158900.1 | - | 3 - 4 | 259 | 112 | ||
| AK168011.1 | - | 3 - 4 | 217 | 112 | ||
| BB848819.1 | - | 3 - 4 | 349 | 112 | ||
| BC012963.1 | - | 4 - 5 | 259 | 4211421 | 112 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ndrg2
Genomic Map