Assay Details
| Gene Symbol: Rhox13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001185002.1 | NP_001171931.1 | 1 - 2 | 978 | 76 | |
| GenBank mRNA | AK007253.1 | - | 1 - 2 | 120 | 76 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | reproductive homeobox 12 | Rhox12 | Gm1148 | -9.9kb | Anti-sense | Chr. X - 38104062 - 38110907 on NCBI Build GRCm38 |
| Downstream | zinc finger and BTB domain containing 33 | Zbtb33 |
AW260253, E130014G12Rik, Kaiso, Znf-kaiso |
66.7kb | Sense | Chr. X - 38189793 - 38197046 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Rhox13
Genomic Map