| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0700030791.mm.1 |
NM_001164086.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030791.mm.1 |
NM_001164086.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030791.mm.1 |
NM_001164087.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030791.mm.1 |
NM_001164087.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030791.mm.1 |
NM_011983.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030791.mm.1 |
NM_011983.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030791.mm.1 |
TC0700003614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030787.mm.1 |
NM_001164086.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030787.mm.1 |
NM_001164086.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030787.mm.1 |
NM_001164087.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030787.mm.1 |
NM_001164087.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030787.mm.1 |
NM_011983.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030787.mm.1 |
NM_011983.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030787.mm.1 |
TC0700003614.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0700030788.mm.1 |
NM_001164087.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030788.mm.1 |
NM_001164087.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030788.mm.1 |
NM_011983.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030788.mm.1 |
NM_011983.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0700030788.mm.1 |
TC0700003614.mm.1 |
MTA TC |