Assay Details
| Gene Symbol: Pde8a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008803.2 | NP_032829.1 | 9 - 10 | 1035 | 124 | |
| GenBank mRNA | AF067806.1 | - | 1101 | 124 | ||
| AK148238.1 | - | 1244 | 124 | |||
| BC125578.1 | - | 9 - 10 | 1035 | 40130093 | 124 | |
| BC132145.1 | - | 9 - 10 | 1035 | 40130422 | 124 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pde8a
Genomic Map