Assay Details
| Gene Symbol: Slc30a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033286.2 | NP_001028458.1 | 1 - 2 | 834 | 59 | |
| GenBank mRNA | AK136932.1 | - | 1 - 2 | 642 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc30a10
Genomic Map