Assay Details
| Gene Symbol: Slc30a10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033286.2 | NP_001028458.1 | 2 - 3 | 913 | 92 | |
| GenBank mRNA | AK053537.1 | - | 1 - 2 | 1588 | 92 | |
| AK136932.1 | - | 2 - 3 | 721 | 92 | ||
| BC054548.1 | - | 1 - 2 | 1543 | 92 | ||
| BC108419.1 | - | 2 - 3 | 301 | 92 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC108419.1 transcripts.
Set Membership: |
|

Slc30a10
Genomic Map