Assay Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_134114.2 | NP_598875.2 | 6 - 7 | 533 | 74 | |
| GenBank mRNA | AK017471.1 | - | 4 - 5 | 277 | 74 | |
| AK087027.1 | - | 1 - 2 | 76 | 74 | ||
| AK151338.1 | - | 6 - 7 | 464 | 74 | ||
| AK171776.1 | - | 4 - 5 | 298 | 74 | ||
| BC006777.1 | - | 5 - 6 | 1923 | 74 | ||
| BC091770.1 | - | 6 - 7 | 534 | 4974098 | 74 | |
| BC130232.1 | - | 6 - 7 | 457 | 40141936 | 74 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Sft2d1
Genomic Map