Assay Details
| Gene Symbol: Sft2d1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK017471.1 | - | 1 - 2 | 96 | 66 | |
| AK171776.1 | - | 1 - 2 | 117 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK171776.1 transcripts.
Set Membership: |
|

Sft2d1
Genomic Map