Assay Details
| Gene Symbol: Fgfr1op | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197046.1 | NP_001183975.1 | 12 - 13 | 1201 | 65 | |
| NM_201230.5 | NP_957682.1 | 11 - 12 | 1141 | 65 | ||
| GenBank mRNA | AK016110.1 | - | 12 - 13 | 1204 | 65 | |
| AK028778.1 | - | 11 - 12 | 1070 | 65 | ||
| AK031261.1 | - | 11 - 12 | 1124 | 65 | ||
| AK050389.1 | - | 11 - 12 | 1079 | 65 | ||
| BC050774.1 | - | 11 - 12 | 1093 | 65 | ||
| BC058804.1 | - | 11 - 12 | 1076 | 65 | ||
| BC080717.1 | - | 12 - 13 | 1188 | 4935625 | 65 | |
| BC108331.1 | - | 12 - 13 | 1199 | 30434232 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of protein kinase activity
positive regulation of cell proliferation
positive regulation of cell growth
positive regulation of cell migration
microtubule anchoring
negative regulation of protein tyrosine kinase activity
protein kinase binding
protein tyrosine kinase inhibitor activity
protein homodimerization activity
negative regulation of protein kinase activity
positive regulation of cell proliferation
positive regulation of cell growth
positive regulation of cell migration
microtubule anchoring
negative regulation of protein tyrosine kinase activity
protein kinase binding
protein tyrosine kinase inhibitor activity
protein homodimerization activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2153827 | BC080717.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2153827 | AK050389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044104 | BC108331.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044104 | BC080717.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044104 | AK050389.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044104 | BC050774.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2044104 | AK031261.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 720557 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 639197 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 316231 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 863891 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 931193 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 67063 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494792 | AK077668.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494792 | AK147133.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494792 | BC025058.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494792 | BC051012.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494792 | BC052920.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494792 | L37092.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494792 | M58633.1 | genbank_mrna_mouse |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 494792 | NM_007661.3 | refseq_mrna_mouse_NM |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 852305 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 637472 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 176031 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 461944 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 445965 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 914103 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 893949 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 40093 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 116621 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 62287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 30037 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P149625 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P164663 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2153827 | BC080717.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2153827 | AK050389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044104 | BC108331.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044104 | BC080717.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044104 | AK050389.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044104 | BC050774.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2044104 | AK031261.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P147778 | BC108331.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P147778 | BC058804.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P147778 | BC080717.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P147778 | AK050389.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_52_P147778 | BC050774.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P417179 | BC108331.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P417179 | BC058804.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P417179 | BC080717.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P417179 | AK050389.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P417179 | BC050774.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09553845 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09553840 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09553831 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P164663 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09553827 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P149625 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09553815 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09553852 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09553821 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P09553834 | - | - |
| Whole Mouse Genome G4122A | A_51_P417179 | BC108331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417179 | BC058804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417179 | BC080717.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417179 | AK050389.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P417179 | BC050774.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P147778 | BC108331.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P147778 | BC058804.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P147778 | BC080717.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P147778 | AK050389.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P147778 | BC050774.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P105201 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P164663 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P149625 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958353 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958348 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958370 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958350 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958369 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958367 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958358 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958365 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958355 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958351 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958366 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958364 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32958349 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10447632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441621 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441630 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441627 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441626 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441624 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441631 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441622 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441632 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441629 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441625 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10447633 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441623 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10441628 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1428919_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10441620 | - | - |
| Mouse Genome 430 2.0 Array | 1428919_at | - | - |
| Mouse Genome 430 2.0 Array | 1428919_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75296_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 75296_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1700000637.mm.1 | NM_001197046.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000637.mm.1 | NM_001197046.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000637.mm.1 | NM_201230.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000637.mm.1 | NM_201230.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000637.mm.1 | TC1700000081.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700000636.mm.1 | NM_001197046.1.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000636.mm.1 | NM_001197046.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000636.mm.1 | NM_201230.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000636.mm.1 | NM_201230.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1700000636.mm.1 | TC1700000081.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR1700000639.mm.1 | TC1700000081.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1428919_PM_at | - | - |
| Mouse Expression Set 430 | 1428919_at | - | - |
| Murine Genome U74v2 Set | 170705_PM_r_at | - | - |
| Murine Genome U74v2 Set | 170705_r_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428919_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1428919_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 6510300 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 105670167 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2696339 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2697790 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2696339 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2697790 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1254742 | - | - |
| mouse-6 v1.1 (Illumina) | scl0001764.1_391-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 105670167 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 105670167 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2696339 | - | - |

Fgfr1op
Genomic Map