Assay Details
| Gene Symbol: Fgfr1op | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197046.1 | NP_001183975.1 | 6 - 7 | 594 | 88 | |
| GenBank mRNA | AK016110.1 | - | 6 - 7 | 597 | 88 | |
| AK029470.1 | - | 6 - 7 | 593 | 88 | ||
| BC080717.1 | - | 6 - 7 | 581 | 4935625 | 88 | |
| BC108331.1 | - | 6 - 7 | 592 | 30434232 | 88 | |
More Information
Set Membership: |
|

Fgfr1op
Genomic Map