Assay Details
| Gene Symbol: Fgfr1op | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197046.1 | NP_001183975.1 | 9 - 10 | 993 | 59 | |
| NM_201230.5 | NP_957682.1 | 8 - 9 | 933 | 59 | ||
| GenBank mRNA | AK016110.1 | - | 9 - 10 | 996 | 59 | |
| AK028778.1 | - | 8 - 9 | 862 | 59 | ||
| AK031261.1 | - | 8 - 9 | 916 | 59 | ||
| AK050389.1 | - | 8 - 9 | 871 | 59 | ||
| BC050774.1 | - | 8 - 9 | 885 | 59 | ||
| BC058804.1 | - | 8 - 9 | 868 | 59 | ||
| BC080717.1 | - | 9 - 10 | 980 | 4935625 | 59 | |
| BC108331.1 | - | 9 - 10 | 991 | 30434232 | 59 | |
More Information
Set Membership: |
|

Fgfr1op
Genomic Map