Assay Details
| Gene Symbol: Fgfr1op | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197046.1 | NP_001183975.1 | 11 - 12 | 1103 | 104 | |
| NM_201230.5 | NP_957682.1 | 10 - 11 | 1043 | 104 | ||
| GenBank mRNA | AK016110.1 | - | 11 - 12 | 1106 | 104 | |
| AK028778.1 | - | 10 - 11 | 972 | 104 | ||
| AK031261.1 | - | 10 - 11 | 1026 | 104 | ||
| AK050389.1 | - | 10 - 11 | 981 | 104 | ||
| BC050774.1 | - | 10 - 11 | 995 | 104 | ||
| BC058804.1 | - | 10 - 11 | 978 | 104 | ||
| BC080717.1 | - | 11 - 12 | 1090 | 4935625 | 104 | |
| BC108331.1 | - | 11 - 12 | 1101 | 30434232 | 104 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fgfr1op
Genomic Map