Assay Details
| Gene Symbol: Mettl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019721.2 | NP_062695.2 | 1 - 2 | 244 | 82 | |
| GenBank mRNA | AK035061.1 | - | 1 - 2 | 238 | 82 | |
| AK085189.1 | - | 1 - 2 | 231 | 82 | ||
| AK151616.1 | - | 1 - 2 | 231 | 82 | ||
| AK153113.1 | - | 1 - 2 | 231 | 82 | ||
| BC012526.1 | - | 1 - 2 | 228 | 3981018 | 82 | |
| BY108113.1 | - | 1 - 2 | 249 | 82 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mettl3
Genomic Map