Assay Details
| Gene Symbol: Samd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC145829.1 | - | 3 - 4 | 1204 | 40131132 | 77 |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Samd4| Gene Symbol: Samd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC145829.1 | - | 3 - 4 | 1204 | 40131132 | 77 |
Set Membership: |
|