Assay Details
| Gene Symbol: Bcam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020486.2 | NP_065232.1 | 10 - 11 | 1381 | 76 | |
| GenBank mRNA | AB035511.1 | - | 10 - 11 | 1342 | 76 | |
| AF109160.1 | - | 10 - 11 | 1381 | 76 | ||
| AF221507.1 | - | 10 - 11 | 1331 | 76 | ||
| AF346663.1 | - | 10 - 11 | 1319 | 76 | ||
| AK075704.1 | - | 10 - 11 | 1346 | 76 | ||
| AK158439.1 | - | 10 - 11 | 1347 | 76 | ||
| BC004826.1 | - | 10 - 11 | 1365 | 2616148 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bcam
Genomic Map