Assay Details
| Gene Symbol: Cdk14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011074.2 | NP_035204.2 | 12 - 13 | 1282 | 75 | |
| GenBank mRNA | AF033655.1 | - | 12 - 13 | 1241 | 75 | |
| AK129226.1 | - | 12 - 13 | 1109 | 75 | ||
| AK138437.1 | - | 11 - 12 | 1523 | 75 | ||
| AK139386.1 | - | 11 - 12 | 1443 | 75 | ||
| AK141535.1 | - | 11 - 12 | 1211 | 75 | ||
| AK165149.1 | - | 11 - 12 | 1127 | 75 | ||
| BC068134.1 | - | 11 - 12 | 1486 | 6834018 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Cdk14
Genomic Map