Assay Details
| Gene Symbol: Thrb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113417.1 | NP_001106888.1 | 5 - 6 | 578 | 115 | |
| NM_009380.3 | NP_033406.1 | 2 - 3 | 664 | 115 | ||
| GenBank mRNA | AB221613.1 | - | 2 - 3 | 431 | 115 | |
| AK133662.1 | - | 2 - 3 | 161 | 115 | ||
| AK158826.1 | - | 5 - 6 | 579 | 115 | ||
| BC089035.1 | - | 2 - 3 | 664 | 30695885 | 115 | |
| BC119552.1 | - | 2 - 3 | 601 | 40057420 | 115 | |
| BC119553.1 | - | 2 - 3 | 605 | 40057421 | 115 | |
| S62756.1 | - | 4 - 5 | 448 | 115 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Thrb
Genomic Map