Assay Details
| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128605.1 | NP_001122077.1 | 7 - 8 | 969 | 91 | |
| NM_011183.3 | NP_035313.2 | 7 - 8 | 978 | 91 | ||
| GenBank mRNA | AF038935.1 | - | 6 - 7 | 906 | 91 | |
| AK148111.1 | - | 7 - 8 | 970 | 91 | ||
| AK154119.1 | - | 7 - 8 | 914 | 91 | ||
| AY864737.1 | - | 2 - 3 | 324 | 91 | ||
| BC010403.1 | - | 6 - 7 | 924 | 4038761 | 91 | |
| BM935552.1 | - | 1 - 2 | 92 | 91 | ||
| U57324.1 | - | 7 - 8 | 929 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psen2
Genomic Map