Assay Details
| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128605.1 | NP_001122077.1 | 9 - 10 | 1152 | 95 | |
| NM_011183.3 | NP_035313.2 | 9 - 10 | 1161 | 95 | ||
| GenBank mRNA | AF038935.1 | - | 8 - 9 | 1086 | 95 | |
| AK148111.1 | - | 9 - 10 | 1153 | 95 | ||
| AK154119.1 | - | 9 - 10 | 1097 | 95 | ||
| AY864737.1 | - | 4 - 5 | 507 | 95 | ||
| BC010403.1 | - | 8 - 9 | 1107 | 4038761 | 95 | |
| BM935552.1 | - | 3 - 4 | 275 | 95 | ||
| U57324.1 | - | 9 - 10 | 1112 | 95 | ||
| U57325.1 | - | 2 - 3 | 304 | 95 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Psen2
Genomic Map