Assay Details
| Gene Symbol: Psen2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001128605.1 | NP_001122077.1 | 1 - 2 | 70 | ||
| GenBank mRNA | AK148111.1 | - | 1 - 2 | 70 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK148111.1, NM_001128605.1 transcripts.
Set Membership: |
|

Psen2
Genomic Map