Assay Details
| Gene Symbol: Ndst3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293682.1 | NP_001280611.1 | 9 - 10 | 1330 | 82 | |
| NM_031186.3 | NP_112463.2 | 10 - 11 | 2443 | 82 | ||
| NR_121616.1 | 11 - 12 | 2547 | 82 | |||
| GenBank mRNA | AK014990.1 | - | 8 - 9 | 1483 | 82 | |
| AK015768.1 | - | 9 - 10 | 1330 | 82 | ||
| AK141945.1 | - | 11 - 12 | 2548 | 82 | ||
| BC079622.1 | - | 10 - 11 | 2314 | 30535990 | 82 | |
More Information
Set Membership: |
|

Ndst3
Genomic Map