Assay Details
| Gene Symbol: Extl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018788.3 | NP_061258.2 | 3 - 4 | 2815 | 81 | |
| GenBank mRNA | AF083550.1 | - | 1 - 2 | 2605 | 81 | |
| AK137842.1 | - | 1 - 2 | 1892 | 81 | ||
| AK220344.1 | - | 1 - 2 | 2158 | 81 | ||
| BC058526.1 | - | 1 - 2 | 2206 | 81 | ||
| BC065073.1 | - | 1 - 2 | 2280 | 6834580 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Extl3
Genomic Map