Assay Details
| Gene Symbol: Ebf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001276387.1 | NP_001263316.1 | 13 - 14 | 2139 | 71 | |
| NM_010095.6 | NP_034225.1 | 13 - 14 | 2134 | 71 | ||
| GenBank mRNA | AK053013.1 | - | 13 - 14 | 2135 | 71 | |
| AK139262.1 | - | 13 - 14 | 1846 | 71 | ||
| BC049188.1 | - | 12 - 13 | 1433 | 6312260 | 71 | |
| BC050922.1 | - | 12 - 13 | 1656 | 6409737 | 71 | |
| U71189.1 | - | 12 - 13 | 1319 | 71 | ||
| U82441.1 | - | 12 - 13 | 1188 | 71 | ||
| U92703.1 | - | 12 - 13 | 1539 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ebf2
Genomic Map