Assay Details
| Gene Symbol: Fpgt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029330.2 | NP_083606.2 | 1 - 2 | 85 | 73 | |
| GenBank mRNA | AJ276067.2 | - | 1 - 2 | 72 | 73 | |
| AK028640.1 | - | 1 - 2 | 82 | 73 | ||
| AK045369.1 | - | 1 - 2 | 86 | 73 | ||
| BC110551.1 | - | 1 - 2 | 85 | 40047377 | 73 | |
| BC110552.1 | - | 1 - 2 | 82 | 40047378 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Fpgt
Genomic Map