Assay Details
| Gene Symbol: Zranb2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_017381.2 | NP_059077.1 | 1 - 2 | 82 | 86 | |
| GenBank mRNA | AF133818.1 | - | 1 - 2 | 60 | 86 | |
| AK146275.1 | - | 1 - 2 | 80 | 86 | ||
| AK146854.1 | - | 1 - 2 | 74 | 86 | ||
| AK150291.1 | - | 1 - 2 | 121 | 86 | ||
| AK152536.1 | - | 1 - 2 | 121 | 86 | ||
| AK160862.1 | - | 1 - 2 | 79 | 86 | ||
| AK166534.1 | - | 1 - 2 | 77 | 86 | ||
| AK167756.1 | - | 1 - 2 | 80 | 86 | ||
| AK167766.1 | - | 1 - 2 | 110 | 86 | ||
| AK167862.1 | - | 1 - 2 | 110 | 86 | ||
| AK167983.1 | - | 1 - 2 | 110 | 86 | ||
| AK168033.1 | - | 1 - 2 | 80 | 86 | ||
| AK168162.1 | - | 1 - 2 | 81 | 86 | ||
| AK168613.1 | - | 1 - 2 | 112 | 86 | ||
| AK169114.1 | - | 1 - 2 | 74 | 86 | ||
| AK169335.1 | - | 1 - 2 | 83 | 86 | ||
| BC049115.1 | - | 1 - 2 | 92 | 86 | ||
| BC132547.1 | - | 1 - 2 | 79 | 40130824 | 86 | |
| BC138575.1 | - | 1 - 2 | 79 | 8861595 | 86 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK160862.1 transcripts.
Set Membership: |
|

Zranb2
Genomic Map