Assay Details
| Gene Symbol: Lamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017959.2 | NP_001017959.1 | 5 - 6 | 914 | 63 | |
| NM_001290485.1 | NP_001277414.1 | 5 - 6 | 914 | 63 | ||
| NM_010685.4 | NP_034815.2 | 5 - 6 | 914 | 63 | ||
| GenBank mRNA | AK012149.1 | - | 5 - 6 | 873 | 63 | |
| AK032974.1 | - | 5 - 6 | 876 | 63 | ||
| AK048174.1 | - | 5 - 6 | 856 | 63 | ||
| AK051989.1 | - | 4 - 5 | 1712 | 63 | ||
| AK078193.1 | - | 5 - 6 | 908 | 63 | ||
| AK088804.1 | - | 6 - 7 | 891 | 63 | ||
| AK128946.1 | - | 3 - 4 | 576 | 63 | ||
| AK136550.1 | - | 5 - 6 | 875 | 63 | ||
| AK146731.1 | - | 5 - 6 | 895 | 63 | ||
| AK150766.1 | - | 5 - 6 | 883 | 63 | ||
| AK159272.1 | - | 5 - 6 | 869 | 63 | ||
| AK159731.1 | - | 5 - 6 | 869 | 63 | ||
| AK163933.1 | - | 5 - 6 | 915 | 63 | ||
| BC138718.1 | - | 5 - 6 | 801 | 8861740 | 63 | |
| BC138723.1 | - | 5 - 6 | 801 | 8861745 | 63 | |
| J05287.1 | - | 5 - 6 | 841 | 63 | ||
| M32017.1 | - | 1 - 2 | 202 | 63 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Lamp2
Genomic Map